Diphenyl(4-iodophenyl)sulphonium triflate

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Iodophenyl)diphenylsulphonium trifluoromethanesulphonate

PC3898-1G
APOSPC3898-1GEA 168 EUR
APOSPC3898-1G
Diphenyl(4-iodophenyl)sulphonium triflate
Diphenyl(4-iodophenyl)sulphonium triflate

Photoinitiator/Photoacid Generator

Formel: C₁₉H₁₄F₃IO₃S₂
Molecular Weight: 538,342 g/mol
Schmelzpunkt: 155…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683574
CAS-Nummer: 255056-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR