2-Brom-6-fluorbenzoesäure

Lieferant: Apollo Scientific

PC3918-5G
APOSPC3918-5GEA 21.9 EUR
APOSPC3918-5G
2-Brom-6-fluorbenzoesäure
2-Bromo-6-fluorobenzoic acid
Formel: C₇H₄BrFO₂
MDL: MFCD01569539
CAS-Nummer: 2252-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR