3,4,5-Trifluor-o-phenylendiamin

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2-Diamino-3,4,5-trifluorbenzol, 3,4,5-Trifluorbenzol-1,2-diamin, 3,4,5-Trifluor-1,2-phenylendiamin

PC3920-1G
APOSPC3920-1GEA 126 EUR
APOSPC3920-1G
3,4,5-Trifluor-o-phenylendiamin
3,4,5-Trifluor-o-phenylendiamin
Formel: C₆H₅F₃N₂
Molecular Weight: 162,11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01569524
CAS-Nummer: 168966-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR