2-Amino-3-brom-5-fluorpyridin

Lieferant: Apollo Scientific
Gefahr

PC3934-1G
APOSPC3934-1GEA 21.9 EUR
APOSPC3934-1G
2-Amino-3-brom-5-fluorpyridin
2-Amino-3-brom-5-fluorpyridin
Formel: C₅H₄BrFN₂
Molecular Weight: 191 g/mol
Schmelzpunkt: 62…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03092924
CAS-Nummer: 869557-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR