3-Brom-5-(trifluormethoxy)benzylbromid 97%

Lieferant: Apollo Scientific
Gefahr

PC3957-10G PC3957-5G PC3957-1G
APOSPC3957-10GEA 0 EUR
APOSPC3957-10G APOSPC3957-5G APOSPC3957-1G
3-Brom-5-(trifluormethoxy)benzylbromid 97%
3-Brom-5-(trifluormethoxy)benzylbromid
Formel: C₈H₅Br₂F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD11035864
CAS-Nummer: 1031929-10-8
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR