4-[(4-Fluorphenyl)ethinyl]phenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-((4-Fluorophenyl)ethynyl)phenol, 1-(4-Fluorophenyl)-2-(4-hydroxyphenyl)acetylene
PC3963-1G
APOSPC3963-1GEA
201
EUR
APOSPC3963-1G
4-[(4-Fluorphenyl)ethinyl]phenol
4-[(4-Fluorphenyl)ethinyl]phenol
Formel:
C₁₄H₉FO Molecular Weight: 212,22 g/mol Schmelzpunkt: 149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093472 CAS-Nummer: 197770-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...