4-Benzyloxy-2-fluorphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Benzyloxy-2-fluorobenzeneboronic acid, (4-(Benzyloxy)-2-fluorophenyl)boronic acid

PC3973-250MG
APOSPC3973-250MGEA 31.9 EUR
APOSPC3973-250MG
4-Benzyloxy-2-fluorphenylboronsäure 97%
4-Benzyloxy-2-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246,04 g/mol
Schmelzpunkt: 151…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093064
CAS-Nummer: 166744-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR