4-Benzyloxy-3-fluorphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Benzyloxy-3-fluorobenzeneboronic acid

PC3974-5G
APOSPC3974-5GEA 65.4 EUR
APOSPC3974-5G
4-Benzyloxy-3-fluorphenylboronsäure 97%
4-Benzyloxy-3-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246.04 g/mol
Schmelzpunkt: 129…131 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00994627
CAS-Nummer: 133057-83-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR