4-Fluor-2-nitrophenol 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC3978-100G
PC3978-25G
PC3978-500G
APOSPC3978-100GEA
53.8
EUR
APOSPC3978-100G
APOSPC3978-25G
APOSPC3978-500G
4-Fluor-2-nitrophenol 99%
4-Fluor-2-nitrophenol
Formel:
C₆H₄FNO₃ Schmelzpunkt: 73…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042528 CAS-Nummer: 394-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...