4-Fluorobenzo[b]furan-7-boronic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Fluorobenzofuran-7-yl)boronic acid, 7-Borono-4-fluorobenzo[b]furan, 4-Fluorobenzofuran-7-boronic acid, 4-Fluoro-1-benzofuran-7-boronic acid
PC400001-1G
APOSPC400001-1GEA
942
EUR
APOSPC400001-1G
4-Fluorobenzo[b]furan-7-boronic acid 95%
4-Fluorobenzo[b]furan-7-boronic acid
Formel:
C₈H₆BFO₃ Molecular Weight: 179,941 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11109423 CAS-Nummer: 1204580-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...