2-(4-(Trifluoromethyl)phenyl)morpholine

Lieferant: Apollo Scientific

PC400017-1G
APOSPC400017-1GEA 945 EUR
APOSPC400017-1G
2-(4-(Trifluoromethyl)phenyl)morpholine
2-(4-(Trifluoromethyl)phenyl)morpholine
Formel: C₁₁H₁₂F₃NO
Molecular Weight: 231,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11646253
CAS-Nummer: 62243-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR