2-(4-(Trifluoromethyl)phenyl)morpholine hydrochloride

Lieferant: Apollo Scientific

PC400018-1G
APOSPC400018-1GEA 2350 EUR
APOSPC400018-1G
2-(4-(Trifluoromethyl)phenyl)morpholine hydrochloride
2-(4-(Trifluoromethyl)phenyl)morpholine hydrochloride
Formel: C₁₁H₁₂F₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD09832156
CAS-Nummer: 1357946-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR