2-[2-(4-Morpholinyl)ethoxy]-5-(trifluoromethyl)aniline

Lieferant: Apollo Scientific

Synonyme: 2-(2-Morpholinoethoxy)-5-(trifluoromethyl)benzenamine, 2-(2-Morpholinoethoxy)-5-(trifluoromethyl)aniline

PC400019-1G
APOSPC400019-1GEA 1830 EUR
APOSPC400019-1G
2-[2-(4-Morpholinyl)ethoxy]-5-(trifluoromethyl)aniline
2-[2-(4-Morpholinyl)ethoxy]-5-(trifluoromethyl)aniline
Formel: C₁₃H₁₇F₃N₂O₂
Molecular Weight: 290,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08688414
CAS-Nummer: 862873-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR