(3-Ethynyl-4-fluorophenoxy)(tert-butyl)dimethylsilane

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl(3-ethynyl-4-fluorophenoxy)dimethylsilane

PC400022-250MG
APOSPC400022-250MGEA 220 EUR
APOSPC400022-250MG
(3-Ethynyl-4-fluorophenoxy)(tert-butyl)dimethylsilane
(3-Ethynyl-4-fluorophenoxy)(tert-butyl)dimethylsilane
Formel: C₁₄H₁₉FOSi
Molecular Weight: 250,384 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536740
CAS-Nummer: 2187435-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR