(6-Fluoro-4-(isopropylamino)pyridin-3-yl)methanol

Lieferant: Apollo Scientific
Achtung

PC400024-250MG
APOSPC400024-250MGEA 387 EUR
APOSPC400024-250MG
(6-Fluoro-4-(isopropylamino)pyridin-3-yl)methanol
(6-Fluoro-4-(isopropylamino)pyridin-3-yl)methanol
Formel: C₉H₁₃FN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD31536742
CAS-Nummer: 2187435-33-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR