(6-Fluoro-4-(methylamino)pyridin-3-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
PC400025-250MG
APOSPC400025-250MGEA
347
EUR
APOSPC400025-250MG
(6-Fluoro-4-(methylamino)pyridin-3-yl)methanol
(6-Fluoro-4-(methylamino)pyridin-3-yl)methanol
Formel:
C₇H₉FN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD31536743 CAS-Nummer: 2187435-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...