1-(2-Bromoethynyl)-4-chloro-2-fluorobenzene

Lieferant: Apollo Scientific
Gefahr

PC400027-250MG
APOSPC400027-250MGEA 192 EUR
APOSPC400027-250MG
1-(2-Bromoethynyl)-4-chloro-2-fluorobenzene
1-(2-Bromoethynyl)-4-chloro-2-fluorobenzene
Formel: C₈H₃BrClF
Lagertemperatur: Raumtemperatur
MDL: MFCD31536745
CAS-Nummer: 2187435-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR