2-(2-Bromoethynyl)-1-chloro-3-fluorobenzene
Lieferant: Apollo Scientific
|
Achtung
|
PC400028-250MG
APOSPC400028-250MGEA
567
EUR
APOSPC400028-250MG
2-(2-Bromoethynyl)-1-chloro-3-fluorobenzene
2-(2-Bromoethynyl)-1-chloro-3-fluorobenzene
Formel:
C₈H₃BrClF Lagertemperatur: Gefrierschrank |
MDL:
MFCD31536746 CAS-Nummer: 2227272-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...