2-(2-Bromoethynyl)-4-chloro-1-fluorobenzene

Lieferant: Apollo Scientific
Achtung

PC400029-250MG
APOSPC400029-250MGEA 567 EUR
APOSPC400029-250MG
2-(2-Bromoethynyl)-4-chloro-1-fluorobenzene
2-(2-Bromoethynyl)-4-chloro-1-fluorobenzene
Formel: C₈H₃BrClF
Lagertemperatur: Raumtemperatur
MDL: MFCD31536747
CAS-Nummer: 2187434-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR