5-(Difluoromethyl)pyridine-2-carbonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
PC400030-500MG
PC400030-250MG
APOSPC400030-500MGEA
0
EUR
APOSPC400030-500MG
APOSPC400030-250MG
5-(Difluoromethyl)pyridine-2-carbonitrile
5-(Difluoromethyl)pyridine-2-carbonitrile
Formel:
C₇H₄F₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18256046 CAS-Nummer: 1211540-57-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...