5-(Difluoromethyl)pyridine-2-carbonitrile

Lieferant: Apollo Scientific
Gefahr

PC400030-500MG PC400030-250MG
APOSPC400030-500MGEA 0 EUR
APOSPC400030-500MG APOSPC400030-250MG
5-(Difluoromethyl)pyridine-2-carbonitrile
5-(Difluoromethyl)pyridine-2-carbonitrile
Formel: C₇H₄F₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18256046
CAS-Nummer: 1211540-57-6
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR