5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde

Lieferant: Apollo Scientific
Achtung

PC400031-1G PC400031-5G
APOSPC400031-1GEA 75.4 EUR
APOSPC400031-1G APOSPC400031-5G
5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde
5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde
Formel: C₁₃H₁₉FO₂Si
Lagertemperatur: Raumtemperatur
MDL: MFCD17676576
CAS-Nummer: 113984-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR