8-Fluoro-2-mercaptoquinazolin-4(3H)-one 95+%

Lieferant: Apollo Scientific
Achtung

PC400045-1G
APOSPC400045-1GEA 191 EUR
APOSPC400045-1G
8-Fluoro-2-mercaptoquinazolin-4(3H)-one 95+%
8-Fluoro-2-mercaptoquinazolin-4(3H)-one
Formel: C₈H₅FN₂OS
Molecular Weight: 196,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561223
CAS-Nummer: 1597549-01-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR