5-Fluoro-2-mercaptoquinazolin-4(3H)-one 95+%

Lieferant: Apollo Scientific
Achtung

PC400049-1G
APOSPC400049-1GEA 192 EUR
APOSPC400049-1G
5-Fluoro-2-mercaptoquinazolin-4(3H)-one 95+%
5-Fluoro-2-mercaptoquinazolin-4(3H)-one
Formel: C₈H₅FN₂OS
Molecular Weight: 196,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561230
CAS-Nummer: 1098336-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR