6,7-Difluoro-2-mercaptoquinazolin-4(3H)-one 95+%

Lieferant: Apollo Scientific
Achtung

PC400050-1G
APOSPC400050-1GEA 325 EUR
APOSPC400050-1G
6,7-Difluoro-2-mercaptoquinazolin-4(3H)-one 95+%
6,7-Difluoro-2-mercaptoquinazolin-4(3H)-one
Formel: C₈H₄F₂N₂OS
Molecular Weight: 214,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561231
CAS-Nummer: 1593461-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR