4-(3-(Trifluoromethyl)-3H-diazirin-3-yl)-2-hydroxybenzaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-4-(3-(trifluoromethyl)-3H-diazirin-3-yl)benzaldehyde

PC400068-250MG
APOSPC400068-250MGEA 536 EUR
APOSPC400068-250MG
4-(3-(Trifluoromethyl)-3H-diazirin-3-yl)-2-hydroxybenzaldehyde
4-(3-(Trifluoromethyl)-3H-diazirin-3-yl)-2-hydroxybenzaldehyde
Formel: C₉H₅F₃N₂O₂
Molecular Weight: 230,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30468740
CAS-Nummer: 308085-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR