5-Bromo-6-(difluoromethoxy)pyridine-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC400106-100MG
PC400106-250MG
APOSPC400106-100MGEA
183
EUR
APOSPC400106-100MG
APOSPC400106-250MG
5-Bromo-6-(difluoromethoxy)pyridine-3-carboxylic acid
5-Bromo-6-(difluoromethoxy)pyridine-3-carboxylic acid
Formel:
C₇H₄BrF₂NO₃ Molecular Weight: 268.012 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31621143 CAS-Nummer: 2231674-35-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...