(5-Bromo-6-(difluoromethoxy)pyridin-3-yl)methanamine

Lieferant: Apollo Scientific
Gefahr

PC400107-100MG PC400107-250MG
APOSPC400107-100MGEA 217 EUR
APOSPC400107-100MG APOSPC400107-250MG
(5-Bromo-6-(difluoromethoxy)pyridin-3-yl)methanamine
(5-Bromo-6-(difluoromethoxy)pyridin-3-yl)methanamine
Formel: C₇H₇BrF₂N₂O
Molecular Weight: 253.044 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621147
CAS-Nummer: 2231675-66-2
UN: 2735
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR