3-Chloro-4-(difluoromethoxy)pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-4-(difluoromethoxy)picolinic acid

PC400113-250MG
APOSPC400113-250MGEA 503 EUR
APOSPC400113-250MG
3-Chloro-4-(difluoromethoxy)pyridine-2-carboxylic acid
3-Chloro-4-(difluoromethoxy)pyridine-2-carboxylic acid
Formel: C₇H₄ClF₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28751019
CAS-Nummer: 1805955-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR