4-(Difluoromethoxy)-3-fluoropyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC400114-250MG PC400114-100MG
APOSPC400114-250MGEA 870 EUR
APOSPC400114-250MG APOSPC400114-100MG
4-(Difluoromethoxy)-3-fluoropyridine-2-carboxylic acid
4-(Difluoromethoxy)-3-fluoropyridine-2-carboxylic acid
Formel: C₇H₄F₃NO₃
Molecular Weight: 207.107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28789683
CAS-Nummer: 1807188-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR