(4-(Difluoromethoxy)pyridin-2-yl)methanol

Lieferant: Apollo Scientific
Achtung

PC400115-250MG
APOSPC400115-250MGEA 304 EUR
APOSPC400115-250MG
(4-(Difluoromethoxy)pyridin-2-yl)methanol
(4-(Difluoromethoxy)pyridin-2-yl)methanol
Formel: C₇H₇F₂NO₂
Molecular Weight: 175,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22552054
CAS-Nummer: 1315361-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR