(5-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanol

Lieferant: Apollo Scientific
Achtung

PC400116-250MG
APOSPC400116-250MGEA 538 EUR
APOSPC400116-250MG
(5-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanol
(5-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanol
Formel: C₇H₆ClF₂NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD28750802
CAS-Nummer: 1805028-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR