(4-(Difluoromethoxy)-5-fluoropyridin-2-yl)methanol

Lieferant: Apollo Scientific
Achtung

PC400117-250MG
APOSPC400117-250MGEA 473 EUR
APOSPC400117-250MG
(4-(Difluoromethoxy)-5-fluoropyridin-2-yl)methanol
(4-(Difluoromethoxy)-5-fluoropyridin-2-yl)methanol
Formel: C₇H₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28789514
CAS-Nummer: 1806275-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR