(3-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanol

Lieferant: Apollo Scientific
Achtung

PC400118-250MG
APOSPC400118-250MGEA 473 EUR
APOSPC400118-250MG
(3-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanol
(3-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanol
Formel: C₇H₆ClF₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28750791
CAS-Nummer: 1805954-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR