(4-(Difluoromethoxy)-3-fluoropyridin-2-yl)methanol

Lieferant: Apollo Scientific
Achtung

PC400119-250MG
APOSPC400119-250MGEA 493 EUR
APOSPC400119-250MG
(4-(Difluoromethoxy)-3-fluoropyridin-2-yl)methanol
(4-(Difluoromethoxy)-3-fluoropyridin-2-yl)methanol
Formel: C₇H₆F₃NO₂
Molecular Weight: 193,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28789512
CAS-Nummer: 1803838-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR