4-(Difluoromethoxy)pyridine-2-carbaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 4-(Difluoromethoxy)picolinaldehyde

PC400120-250MG
APOSPC400120-250MGEA 873 EUR
APOSPC400120-250MG
4-(Difluoromethoxy)pyridine-2-carbaldehyde 95%
4-(Difluoromethoxy)pyridine-2-carbaldehyde
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22552055
CAS-Nummer: 1315361-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR