tert-Butyl (5-chloro-4-(difluoromethoxy)pyridin-2-yl)methylcarbamate

Lieferant: Apollo Scientific
Achtung

PC400125-250MG
APOSPC400125-250MGEA 369 EUR
APOSPC400125-250MG
tert-Butyl (5-chloro-4-(difluoromethoxy)pyridin-2-yl)methylcarbamate
tert-Butyl (5-chloro-4-(difluoromethoxy)pyridin-2-yl)methylcarbamate
Formel: C₁₂H₁₅ClF₂N₂O₃
Molecular Weight: 308,709 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621171
CAS-Nummer: 2231675-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR