(4-(Difluoromethoxy)pyridin-2-yl)methanamine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

PC400127-250MG
APOSPC400127-250MGEA 696 EUR
APOSPC400127-250MG
(4-(Difluoromethoxy)pyridin-2-yl)methanamine hydrochloride 95%
(4-(Difluoromethoxy)pyridin-2-yl)methanamine hydrochloride
Formel: C₇H₈F₂N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD31621175
CAS-Nummer: 2231673-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR