(5-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC400128-250MG
APOSPC400128-250MGEA 462 EUR
APOSPC400128-250MG
(5-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanamine hydrochloride
(5-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanamine hydrochloride
Formel: C₇H₇ClF₂N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD31621177
CAS-Nummer: 2231676-84-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR