(4-(Difluoromethoxy)-5-fluoropyridin-2-yl)methanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC400129-250MG
APOSPC400129-250MGEA 1120 EUR
APOSPC400129-250MG
(4-(Difluoromethoxy)-5-fluoropyridin-2-yl)methanamine hydrochloride
(4-(Difluoromethoxy)-5-fluoropyridin-2-yl)methanamine hydrochloride
Formel: C₇H₇F₃N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD31621179
CAS-Nummer: 2231675-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR