(3-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC400130-250MG
APOSPC400130-250MGEA 462 EUR
APOSPC400130-250MG
(3-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanamine hydrochloride
(3-Chloro-4-(difluoromethoxy)pyridin-2-yl)methanamine hydrochloride
Formel: C₇H₇ClF₂N₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD31621181
CAS-Nummer: 2231674-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR