(4-(Difluoromethoxy)-3-fluoropyridin-2-yl)methanamine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
PC400131-250MG
APOSPC400131-250MGEA
406
EUR
APOSPC400131-250MG
(4-(Difluoromethoxy)-3-fluoropyridin-2-yl)methanamine hydrochloride
(4-(Difluoromethoxy)-3-fluoropyridin-2-yl)methanamine hydrochloride
Formel:
C₇H₇F₃N₂O·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD31621183 CAS-Nummer: 2231673-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...