4-(Difluoromethoxy)pyridin-2-amine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC400137-250MG
APOSPC400137-250MGEA
125
EUR
APOSPC400137-250MG
4-(Difluoromethoxy)pyridin-2-amine 95%
4-(Difluoromethoxy)pyridin-2-amine
Formel:
C₆H₆F₂N₂O Molecular Weight: 160,121 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22550528 CAS-Nummer: 1206972-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...