3-Chloro-4-(difluoromethoxy)pyridin-2-amine

Lieferant: Apollo Scientific
Achtung

PC400140-250MG
APOSPC400140-250MGEA 725 EUR
APOSPC400140-250MG
3-Chloro-4-(difluoromethoxy)pyridin-2-amine
3-Chloro-4-(difluoromethoxy)pyridin-2-amine
Formel: C₆H₅ClF₂N₂O
Molecular Weight: 194,567 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621124
CAS-Nummer: 2231676-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR