(2-(Difluoromethoxy)pyridin-3-yl)methanol

Lieferant: Apollo Scientific
Achtung

PC400159-250MG
APOSPC400159-250MGEA 609 EUR
APOSPC400159-250MG
(2-(Difluoromethoxy)pyridin-3-yl)methanol
(2-(Difluoromethoxy)pyridin-3-yl)methanol
Formel: C₇H₇F₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31621140
CAS-Nummer: 2222104-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR