2-(Difluoromethoxy)pyridine-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Difluoromethoxy)nicotinaldehyde

PC400160-250MG
APOSPC400160-250MGEA 416 EUR
APOSPC400160-250MG
2-(Difluoromethoxy)pyridine-3-carbaldehyde
2-(Difluoromethoxy)pyridine-3-carbaldehyde
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621142
CAS-Nummer: 2222104-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR