5-Bromo-6-fluoroisoquinoline

Lieferant: Apollo Scientific
Achtung

PC400167-1G PC400167-250MG
APOSPC400167-1GEA 329 EUR
APOSPC400167-1G APOSPC400167-250MG
5-Bromo-6-fluoroisoquinoline
5-Bromo-6-fluoroisoquinoline
Formel: C₉H₅BrFN
Molecular Weight: 226.045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20527917
CAS-Nummer: 1239463-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR