2-Bromo-3-fluoro-5-(methylthio)pyridine

Lieferant: Apollo Scientific
Achtung

PC400175-1G PC400175-250MG
APOSPC400175-1GEA 340 EUR
APOSPC400175-1G APOSPC400175-250MG
2-Bromo-3-fluoro-5-(methylthio)pyridine
2-Bromo-3-fluoro-5-(methylthio)pyridine
Formel: C₆H₅BrFNS
Molecular Weight: 222.078 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26127752
CAS-Nummer: 1820608-42-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR