6-Bromo-5-fluoropicolinonitrile

Lieferant: Apollo Scientific
Achtung

PC400176-250MG
APOSPC400176-250MGEA 270 EUR
APOSPC400176-250MG
6-Bromo-5-fluoropicolinonitrile
6-Bromo-5-fluoropicolinonitrile
Formel: C₆H₂BrFN₂
Molecular Weight: 200,996 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22684691
CAS-Nummer: 1416713-45-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR