4-Ethynyl-3-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC400177-1G PC400177-250MG
APOSPC400177-1GEA 239 EUR
APOSPC400177-1G APOSPC400177-250MG
4-Ethynyl-3-fluoroaniline
4-Ethynyl-3-fluoroaniline
Formel: C₈H₆FN
Lagertemperatur: Raumtemperatur
MDL: MFCD11036455
CAS-Nummer: 1233501-57-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR